Imprimer

Anti-NOS2 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15326-7EA 586 EUR
ABCAAB15326-7
Anti-NOS2 Rabbit polyclonal antibody
Type: Primary
Antigen: NOS2 (nitric oxide synthase 2, inducible)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR