Imprimer

Anti-PSEN1 Mouse Monoclonal Antibody [clone: APS 18]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15458-200EA 907 EUR
ABCAAB15458-200
Anti-PSEN1 Mouse Monoclonal Antibody [clone: APS 18]
Anticorps
Type: Primary
Antigen: PSEN1 (presenilin 1)
Clonality: monoclonal
Clone: APS 18
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR