Imprimer

Anti-ABCC4 Rat Monoclonal Antibody [clone: M4I-10]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15602-500EA 720 EUR
ABCAAB15602-500
Anti-ABCC4 Rat Monoclonal Antibody [clone: M4I-10]
Anticorps
Type: Primary
Antigen: ABCC4 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4)
Clonality: monoclonal
Clone: M4I-10
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR