Imprimer

Anti-Ly6c Rat Monoclonal Antibody [clone: ER-MP20]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15627-50EA 742 EUR
ABCAAB15627-50
Anti-Ly6c Rat Monoclonal Antibody [clone: ER-MP20]
Anticorps
Type: Primary
Antigen: Ly6c
Clonality: monoclonal
Clone: ER-MP20
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR