Imprimer

Anti-Hematopoiesis related Macrophage Rat Monoclonal Antibody [clone: ER-HR3]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15636-250EA 666 EUR
ABCAAB15636-250
Anti-Hematopoiesis related Macrophage Rat Monoclonal Antibody [clone: ER-HR3]
Anticorps
Type: Primary
Antigen: Hematopoiesis related Macrophage
Clonality: monoclonal
Clone: ER-HR3
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2c
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR