Imprimer

Anti-BMP6 Mouse Monoclonal Antibody [clone: morph-6.1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15640-50EA 743 EUR
ABCAAB15640-50
Anti-BMP6 Mouse Monoclonal Antibody [clone: morph-6.1]
Anticorps
Type: Primary
Antigen: BMP6 (bone morphogenetic protein 6)
Clonality: monoclonal
Clone: morph-6.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR