Imprimer

Anti-AIF1 Mouse Monoclonal Antibody [clone: 1022-5]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15690-100EA 739 EUR
ABCAAB15690-100
Anti-AIF1 Mouse Monoclonal Antibody [clone: 1022-5]
Anticorps
Type: Primary
Antigen: AIF1 (allograft inflammatory factor 1)
Clonality: monoclonal
Clone: 1022-5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR