Imprimer

Anti-AIF1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 1022-5]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15691-100EA 885 EUR
ABCAAB15691-100
Anti-AIF1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 1022-5]
Anticorps
Type: Primary
Antigen: AIF1 (allograft inflammatory factor 1)
Clonality: monoclonal
Clone: 1022-5
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR