Imprimer

Anti-UBE2F Goat Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15707-100EA 721 EUR
ABCAAB15707-100
Anti-UBE2F Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: UBE2F (ubiquitin-conjugating enzyme E2F (putative))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR