Imprimer

Anti-SNX9 Goat polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15709-100EA 361 EUR
ABCAAB15709-100
Anti-SNX9 Goat polyclonal antibody
Type: Primary
Antigen: SNX9 (sorting nexin 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR