Imprimer

Anti-PAX3 Goat Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15717-100EA 924 EUR
ABCAAB15717-100
Anti-PAX3 Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PAX3 (paired box 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR