Imprimer

Anti-Staphylococcus TSST1 Sheep polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB15936-500EA 565 EUR
ABCAAB15936-500
Anti-Staphylococcus TSST1 Sheep polyclonal antibody
Type: Primary
Antigen: Staphylococcus TSST1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR