Imprimer

Anti-HTR2A Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16028-100EA 565 EUR
ABCAAB16028-100
Anti-HTR2A Rabbit polyclonal antibody
Type: Primary
Antigen: HTR2A (5-hydroxytryptamine (serotonin) receptor 2A, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR