Imprimer

Anti-TNFSF13B Rat Monoclonal Antibody [clone: Buffy 2]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16081-50EA 723 EUR
ABCAAB16081-50
Anti-TNFSF13B Rat Monoclonal Antibody [clone: Buffy 2]
Anticorps
Type: Primary
Antigen: TNFSF13B (tumor necrosis factor (ligand) superfamily, member 13b)
Clonality: monoclonal
Clone: Buffy 2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR