Imprimer

Anti-TNFSF13 Rat monoclonal antibody [clone: Sacha-1]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16085-50EA 725 EUR
ABCAAB16085-50
Anti-TNFSF13 Rat monoclonal antibody [clone: Sacha-1]
Type: Primary
Antigen: TNFSF13 (tumor necrosis factor (ligand) superfamily, member 13)
Clonality: monoclonal
Clone: Sacha-1
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR