Imprimer

Anti-CHEK1 Sheep Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16130-250EA 704 EUR
ABCAAB16130-250
Anti-CHEK1 Sheep Polyclonal Antibody
Anticorps
Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR