Imprimer

Anti-IL6 Mouse monoclonal antibody Biotin [clone: AS16]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16164-100EA 603 EUR
ABCAAB16164-100
Anti-IL6 Mouse monoclonal antibody Biotin [clone: AS16]
Type: Primary
Antigen: IL6 (interleukin 6 (interferon, beta 2))
Clonality: monoclonal
Clone: AS16
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR