Imprimer

Anti-BAFF Receptor Mouse monoclonal antibody unconjugated [clone: 11C1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16232-100EA 724 EUR
ABCAAB16232-100
Anti-BAFF Receptor Mouse monoclonal antibody unconjugated [clone: 11C1]
Anticorps
Type: Primary
Antigen: BAFF Receptor
Clonality: monoclonal
Clone: 11C1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR