Imprimer

Anti-NR2E3 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16243-100EA 838 EUR
ABCAAB16243-100
Anti-NR2E3 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: NR2E3 (nuclear receptor subfamily 2, group E, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR