Imprimer
Anti-NKAP Chicken Polyclonal Antibody
Fournisseur: Abcam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ABCAAB16295-50EA 691 EUR
ABCAAB16295-50
Anti-NKAP Chicken Polyclonal Antibody
Anticorps
Type: Primary
Antigen: NKAP (NFKB activating protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human