Imprimer

Anti-PCCA Chicken Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16307-50EA 778 EUR
ABCAAB16307-50
Anti-PCCA Chicken Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PCCA (propionyl CoA carboxylase, alpha polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR