Imprimer

Anti-TP53 Mouse Monoclonal Antibody [clone: X77]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16465-100EA 724 EUR
ABCAAB16465-100
Anti-TP53 Mouse Monoclonal Antibody [clone: X77]
Anticorps
Type: Primary
Antigen: TP53 (tumor protein p53)
Clonality: monoclonal
Clone: X77
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR