Imprimer

Anti-DLG4 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16493-100EA 572 EUR
ABCAAB16493-100
Anti-DLG4 Rabbit polyclonal antibody
Type: Primary
Antigen: DLG4 (discs, large homolog 4 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR