Imprimer

Anti-Lambda Light chain Mouse monoclonal antibody [clone: ICO106]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16567-100EA 572 EUR
ABCAAB16567-100
Anti-Lambda Light chain Mouse monoclonal antibody [clone: ICO106]
Type: Primary
Antigen: Lambda Light chain
Clonality: monoclonal
Clone: ICO106
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR