Imprimer

Anti-Cytokeratin 5 + 14 Mouse Monoclonal Antibody [clone: 16.4]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16570-100EA 701 EUR
ABCAAB16570-100
Anti-Cytokeratin 5 + 14 Mouse Monoclonal Antibody [clone: 16.4]
Anticorps
Type: Primary
Antigen: Cytokeratin 5 + 14
Clonality: monoclonal
Clone: 16.4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR