Imprimer

Anti-CCR3 Mouse Monoclonal Antibody (Biotin) [clone: 5E8]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16581-100EA 698 EUR
ABCAAB16581-100
Anti-CCR3 Mouse Monoclonal Antibody (Biotin) [clone: 5E8]
Anticorps
Type: Primary
Antigen: CCR3 (chemokine (C-C motif) receptor 3)
Clonality: monoclonal
Clone: 500000000
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR