Imprimer

Anti-CD44 Mouse Monoclonal Antibody [clone: 156-3C11]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16728-500EA 735 EUR
ABCAAB16728-500
Anti-CD44 Mouse Monoclonal Antibody [clone: 156-3C11]
Anticorps
Type: Primary
Antigen: CD44 (CD44 molecule (Indian blood group))
Clonality: monoclonal
Clone: 156-3C11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR