Imprimer

Anti-PRDX1 Mouse monoclonal antibody [clone: 2A4]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16746-100EA 603 EUR
ABCAAB16746-100
Anti-PRDX1 Mouse monoclonal antibody [clone: 2A4]
Type: Primary
Antigen: PRDX1 (peroxiredoxin 1)
Clonality: monoclonal
Clone: 2A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR