Imprimer

Anti-PRDX3 Mouse monoclonal antibody [clone: 12B]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16751-100EA 621 EUR
ABCAAB16751-100
Anti-PRDX3 Mouse monoclonal antibody [clone: 12B]
Type: Primary
Antigen: PRDX3 (peroxiredoxin 3)
Clonality: monoclonal
Clone: 12B
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR