Imprimer

Anti-H3F3AP6 Mouse monoclonal antibody [clone: EA10]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16767-100EA 621 EUR
ABCAAB16767-100
Anti-H3F3AP6 Mouse monoclonal antibody [clone: EA10]
Type: Primary
Antigen: H3F3AP6 (H3 histone, family 3A, pseudogene 6)
Clonality: monoclonal
Clone: EA10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR