Imprimer

Anti-TXNRD1 Rabbit Polyclonal Antibody

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16840-100EA 751 EUR
ABCAAB16840-100
Anti-TXNRD1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TXNRD1 (thioredoxin reductase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR