Imprimer

Anti-TXNRD1 Mouse Monoclonal Antibody [clone: 19A1]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16847-100EA 714 EUR
ABCAAB16847-100
Anti-TXNRD1 Mouse Monoclonal Antibody [clone: 19A1]
Anticorps
Type: Primary
Antigen: TXNRD1 (thioredoxin reductase 1)
Clonality: monoclonal
Clone: 19A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR