Imprimer

Anti-EMR1 Rat monoclonal antibody unconjugated [clone: BM8]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB16911-100EA 749 EUR
ABCAAB16911-100
Anti-EMR1 Rat monoclonal antibody unconjugated [clone: BM8]
Anticorps
Type: Primary
Antigen: EMR1 (egf-like module containing, mucin-like, hormone receptor-like 1)
Clonality: monoclonal
Clone: BM8
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR