Imprimer

Anti-HIST4H4 Mouse monoclonal antibody unconjugated [clone: mAbcam 17036]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17036-100EA 903 EUR
ABCAAB17036-100
Anti-HIST4H4 Mouse monoclonal antibody unconjugated [clone: mAbcam 17036]
Anticorps
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: monoclonal
Clone: mAbcam 17036
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR