Imprimer

Anti-NUF2 Mouse monoclonal antibody [clone: 28-37]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17058-100EA 540 EUR
ABCAAB17058-100
Anti-NUF2 Mouse monoclonal antibody [clone: 28-37]
Type: Primary
Antigen: NUF2 (NUF2, NDC80 kinetochore complex component)
Clonality: monoclonal
Clone: 28-37
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR