Imprimer

Anti-KRT18 Mouse monoclonal antibody [clone: CY-90]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17070-6EA 606 EUR
ABCAAB17070-6
Anti-KRT18 Mouse monoclonal antibody [clone: CY-90]
Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: monoclonal
Clone: CY-90
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR