Imprimer

Anti-IgM Mouse Monoclonal Antibody [clone: R1/69]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17104-1EA 704 EUR
ABCAAB17104-1
Anti-IgM Mouse Monoclonal Antibody [clone: R1/69]
Anticorps
Type: Primary
Antigen: IgM
Clonality: Monoclonal
Clone: R1/69
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR