Imprimer

Anti-ALK Mouse Monoclonal Antibody [clone: 5A4]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17127-500EA 737 EUR
ABCAAB17127-500
Anti-ALK Mouse Monoclonal Antibody [clone: 5A4]
Anticorps
Type: Primary
Antigen: ALK (anaplastic lymphoma receptor tyrosine kinase)
Clonality: monoclonal
Clone: 5A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR