Imprimer

Anti-ACTB Mouse monoclonal antibody [clone: 1A4+5C5]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17146-500EA 579 EUR
ABCAAB17146-500
Anti-ACTB Mouse monoclonal antibody [clone: 1A4+5C5]
Type: Primary
Antigen: ACTB (actin, beta)
Clonality: monoclonal
Clone: 1A4+5C5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR