Imprimer

Anti-pan Cytokeratin Mouse monoclonal antibody [clone: Lu-5]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17155-1EA 540 EUR
ABCAAB17155-1
Anti-pan Cytokeratin Mouse monoclonal antibody [clone: Lu-5]
Type: Primary
Antigen: pan Cytokeratin
Clonality: monoclonal
Clone: Lu-5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR