Imprimer

Anti-Hirudin Mouse Monoclonal Antibody [clone: 2D7]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17198-100EA 690 EUR
ABCAAB17198-100
Anti-Hirudin Mouse Monoclonal Antibody [clone: 2D7]
Anticorps
Type: Primary
Antigen: Hirudin
Clonality: monoclonal
Clone: 2D7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR