Imprimer

Anti-BRCA1 Mouse monoclonal antibody unconjugated [clone: GLK-2]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17251-250EA 877 EUR
ABCAAB17251-250
Anti-BRCA1 Mouse monoclonal antibody unconjugated [clone: GLK-2]
Anticorps
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: GLK-2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR