Imprimer

Anti-Bone Alkaline Phosphatase Mouse monoclonal antibody [clone: 0.G.2]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17272-100EA 579 EUR
ABCAAB17272-100
Anti-Bone Alkaline Phosphatase Mouse monoclonal antibody [clone: 0.G.2]
Type: Primary
Antigen: Bone Alkaline Phosphatase
Clonality: monoclonal
Clone: 0.G.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR