Imprimer

Anti-Phosphotyrosine Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 3G247]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17305-50EA 572 EUR
ABCAAB17305-50
Anti-Phosphotyrosine Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 3G247]
Type: Primary
Antigen: Phosphotyrosine
Clonality: monoclonal
Clone: 3G247
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR