Imprimer

Anti-LDL (MDA oxidized) Mouse Monoclonal Antibody [clone: 4C7]

Fournisseur: Abcam
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17354-100EA 698 EUR
ABCAAB17354-100
Anti-LDL (MDA oxidized) Mouse Monoclonal Antibody [clone: 4C7]
Anticorps
Type: Primary
Antigen: LDL (MDA oxidized)
Clonality: monoclonal
Clone: 4C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR