Imprimer

Anti-TULP1 Rabbit polyclonal antibody unconjugated

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17364-50EA 0 EUR
ABCAAB17364-50
Anti-TULP1 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: TULP1 (tubby like protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR