Imprimer

Anti-Phosphotyrosine Mouse monoclonal antibody FITC (Fluorescein) [clone: 1.BB.49]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17372-125EA 572 EUR
ABCAAB17372-125
Anti-Phosphotyrosine Mouse monoclonal antibody FITC (Fluorescein) [clone: 1.BB.49]
Type: Primary
Antigen: Phosphotyrosine
Clonality: monoclonal
Clone: 1.BB.49
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR