Imprimer
Anti-BMP2 Rabbit polyclonal antibody Biotin
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ABCAAB17597-50EA 572 EUR
ABCAAB17597-50
Anti-BMP2 Rabbit polyclonal antibody Biotin
Type: Primary
Antigen: BMP2 (bone morphogenetic protein 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human