Imprimer

Anti-TNFRSF13B Goat polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17620-50EA 572 EUR
ABCAAB17620-50
Anti-TNFRSF13B Goat polyclonal antibody
Type: Primary
Antigen: TNFRSF13B (tumor necrosis factor receptor superfamily, member 13B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR