Imprimer

Anti-CXCL1 Rabbit polyclonal antibody Biotin

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

ABCAAB17678-25EA 572 EUR
ABCAAB17678-25
Anti-CXCL1 Rabbit polyclonal antibody Biotin
Type: Primary
Antigen: CXCL1 (chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR